missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ VEGFA (Human) Recombinant Protein

Product Code. 16212290
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µg
50µg
This item is not returnable. View return policy

Product Code. 16212290

Brand: Abnova™ P5816.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human VEGFA (165 a.a.) full-length recombinant protein with His tag expressed in Barley grain (Hordeum vulgare).

Specifications

Accession Number NP_001165097
Concentration 100 μg/mL
Gene ID (Entrez) 7422
Name vascular endothelial growth factor A
Preparation Method Barley grain (Hordeum vulgare) expression system
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterile water to a concentration of no less than 100 ug/mL, store at -20°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Aliquot to avoid repeated freezing and thawing.
Gene Alias MGC70609, VEGF, VEGF-A, VPF
Gene Symbol VEGFA
Species Plants
Protein Tag His
Buffer Lyophilized from PBS, pH 7.2
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.