missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VEGF Receptor 2 Recombinant Rabbit Monoclonal Antibody (001), APC

Rabbit Recombinant Monoclonal Antibody
£107.00 - £332.00
Specifications
| Antigen | VEGF Receptor 2 |
|---|---|
| Clone | 001 |
| Concentration | 0.1 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30535266
![]() |
Invitrogen™
MA560502 |
100 Tests |
£332.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30534591
|
Invitrogen™
MA560503 |
25 Tests |
£107.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF Receptor 2 Recombinant Monoclonal Antibody for Flow
KDR (FLK1, VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors. FLK1 (VEGF Receptor 2) encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of FLK1 (VEGF Receptor 2) are implicated in infantile capillary hemangiomas.Specifications
| VEGF Receptor 2 | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| RUO | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| P35918 | |
| 16542 | |
| Recombinant Mouse VEGFR2/Flk-1/CD309/KDR protein (Met1-Glu762). | |
| Antibody |
| 001 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| KDR | |
| 6130401C07; CD309; CD309 antigen; fetal liver kinase 1; fetal liver kinase-1; FLK1; Flk-1; FLK1 kinase insert domain receptor (a type III receptor tyrosine kinase) (VEGF receptor 2); FLK1 kinase insert domain receptor (VEGF receptor 2); flk-1 type VEGF receptor; KDR; kinase insert domain protein receptor; kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); kinase NYK; Krd-1; Ly73; protein-tyrosine kinase receptor flk-1; sCD309; soluble CD309; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 2; soluble VEGFR2; sVEGFR-2; tyrosine kinase growth factor receptor; vascular endotheial growth factor 2; vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; VEGF R2; VEGF receptor-2; VEGFR; VEGFR2; VEGFR-2 | |
| KDR | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title