Learn More
Invitrogen™ VEGF-164 Monoclonal Antibody (123704)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523769
Description
This antibody shows 50-100% cross-reactivity with recombinant mouse (rm) VEGF120, rmVEGF164, recombinant human (rh) VEGF121, rhVEGF165, and rhVEGF189 and no cross-reactivity with rmVEGF115. This antibody recognizes an epitope found between amino acids 78-120 of rmVEGF120. This segment is present in all VEGF isoforms except rmVEGF115. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
VEGF-164 is a gene target that encodes for the vascular endothelial growth factor. This protein is responsible for promoting angiogenesis. VEGF-164 has been shown to be involved in the development of various diseases, including cancer, cardiovascular disease, and diabetic retinopathy. Targeting this gene has become an important strategy in the development of new therapeutics for these conditions.
Specifications
| VEGF-164 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P16612 | |
| VEGFA | |
| Mouse myeloma cell line NS0-derived recombinant rat VEGF164 Ala27-Arg190. | |
| 500 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| 123704 | |
| Unconjugated | |
| VEGFA | |
| Vascular Endothelial Growth Factor, Vascular Permeability Factor, VEGFA, VPF | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 83785 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.