missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-C Antibody (MM0006-2E65), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60971F
This item is not returnable.
View return policy
Description
VEGF-C Monoclonal specifically detects VEGF-C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| VEGF-C | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
| 7424 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0006-2E65 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| P49767 | |
| VEGFC | |
| Human recombinant VEGF-C | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity is shown with human VEGFA, VEGF-B and VEGF-D. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction