missing translation for 'onlineSavingsMsg'
Learn More

VEGF-C Antibody (9E7), DyLight 594, Novus Biologicals™

Product Code. 18424124 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424124 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424124 Supplier Novus Biologicals Supplier No. NBP118627DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGF-C Monoclonal antibody specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF-C
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 9E7
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Host Species Mouse
Immunogen Recombinant human VEGF-C
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7424
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.