missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-B Antibody (RM0008-6E72), Janelia Fluor™ 549, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-61024JF549
This item is not returnable.
View return policy
Description
VEGF-B Monoclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| VEGF-B | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VEGFB | |
| Mouse recombinant VEGF-B | |
| 0.1 mL | |
| Primary | |
| This antibody recognizes mouse VEGF-B 186 but not 167 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0008-6E72 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
| Rat | |
| Protein G purified | |
| Angiogenesis, Cell Cycle and Replication | |
| 7423 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction