missing translation for 'onlineSavingsMsg'
Learn More

VEGF-B Antibody [FITC], Novus Biologicals™

Product Code. 30732965 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732965 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732965 Supplier Novus Biologicals Supplier No. AF751F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

VEGF-B Polyclonal antibody specifically detects VEGF-B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF-B
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL
Host Species Goat
Immunogen E. coli-derived recombinant human VEGF-B186
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7423
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.