missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-B Antibody (CSL346), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28237CL1
This item is not returnable.
View return policy
Description
VEGF-B Monoclonal antibody specifically detects VEGF-B in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| VEGF-B | |
| Monoclonal | |
| CoraFluor 1 | |
| vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
| VEGFB | |
| 0.1 mL | |
| Angiogenesis, Cell Cycle and Replication | |
| 7423 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| CSL346 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction