missing translation for 'onlineSavingsMsg'
Learn More

VEGF Antibody (26503R) [DyLight 350], Novus Biologicals™

Product Code. 30734701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734701 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734701 Supplier Novus Biologicals Supplier No. FAB293RD

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGF Monoclonal antibody specifically detects VEGF in Human,Primate samples. It is validated for Immunohistochemistry,Western Blot,ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF
Applications Immunohistochemistry, Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 26503R
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGF165, Ala27-Arg191, Accession # NP_001165097
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Cytokines and Growth Factors, Developmental Biology, Glycobiology, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 7422
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.