missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VEGF 165 Monoclonal antibody specifically detects VEGF 165 in Human, Cynomolgus Monkey samples. It is validated for ELISA
Specifications
Specifications
| Antigen | VEGF 165 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 105 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human/Cynomolgus VEGF 165 (Uniprot#: P15692-4; Met 1-Arg 191). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?