missing translation for 'onlineSavingsMsg'
Learn More

VEGF 165 Antibody (105), DyLight 650, Novus Biologicals™

Product Code. 18419078 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419078 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419078 Supplier Novus Biologicals Supplier No. NBP289780C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

VEGF 165 Monoclonal antibody specifically detects VEGF 165 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF 165
Applications ELISA
Classification Monoclonal
Clone 105
Conjugate DyLight 650
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF 165 (P15692-4; Met 1-Arg 191).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 7422
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.