missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VEGF 121 Monoclonal antibody specifically detects VEGF 121 in Human samples. It is validated for ELISA, ELISA
Specifications
Specifications
| Antigen | VEGF 121 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 145 |
| Conjugate | Janelia Fluor 549 |
| Dilution | ELISA, Sandwich ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF 121 (P15692-9; Met1-Arg147). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?