missing translation for 'onlineSavingsMsg'
Learn More

Vecuronium bromide, MedChemExpress

Product Code. 16454247
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16454247

Brand: Medchem Express HYB0118A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vecuronium (ORG NC 45) bromide is a non-depolarizing neuromuscular blocking agent that also acts as a nicotinic acetylcholine receptor (nAChR) inhibitor, a muscle relaxant, and can be used for pre-surgical anesthesia.

Vecuronium (ORG NC 45) bromide is a non-depolarizing neuromuscular blocking agent that also acts as a nicotinic acetylcholine receptor (nAChR) inhibitor, a muscle relaxant, and can be used for pre-surgical anesthesia.

Specifications

Chemical Name or Material Vecuronium bromide
Physical Form Solid
CAS 50700-72-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C34H57BrN2O4
For Use With (Application) Neuroscience-Neuromodulation
Synonym ORG NC 45
Solubility Information DMSO : ≥ 46 mg/mL (72.13 mM)
SMILES C[C@@]1([C@H]2OC(C)=O)[C@](C[C@@H]2[N+]3(CCCCC3)C)([H])[C@@](CC[C@]4([H])[C@@]5(C[C@H](N6CCCCC6)[C@@H](OC(C)=O)C4)C)([H])[C@]5([H])CC1.[Br-]
Molecular Weight (g/mol) 637.73
Formula Weight 637.73
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.