missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ VECTASTAIN(R) ABC anti-Rat IgG AP Immunodetection Kit

Product Code. 18354718 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354718 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354718 Supplier Novus Biologicals™ Supplier No. AK5004NB

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Immunodetection Kit

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. Do not freeze.
For Use With (Application) Western Blot, ELISA, Immunohistochemistry (IHC), Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen), ISH, ELISpot
Species Rat
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.