missing translation for 'onlineSavingsMsg'
Learn More

VE-Statin/EGFL7 Antibody (362907) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30738113 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30738113 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738113 Supplier Novus Biologicals Supplier No. FAB3089MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

VE-Statin/EGFL7 Monoclonal antibody specifically detects VE-Statin/EGFL7 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen VE-Statin/EGFL7
Applications Western Blot
Classification Monoclonal
Clone 362907
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VE-STATIN, Zneu1
Host Species Rat
Immunogen E. coli-derived recombinant mouse VE-Statin Isoform 1, Thr21-Leu275, Accession # Q9QXT5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 51162
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.