missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Statin/EGFL7 Antibody (362907) [Janelia Fluor™ 646], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3089J
This item is not returnable.
View return policy
Description
VE-Statin/EGFL7 Monoclonal antibody specifically detects VE-Statin/EGFL7 in Mouse samples. It is validated for Western Blot
Specifications
| VE-Statin/EGFL7 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VE-STATIN, Zneu1 | |
| E. coli-derived recombinant mouse VE-Statin Isoform 1, Thr21-Leu275, Accession # Q9QXT5 | |
| 0.1 mL | |
| Angiogenesis | |
| 51162 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot | |
| 362907 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction