missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VE-cadherin Recombinant Rabbit Monoclonal Antibody (9W5E3)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557860

 View more versions of this product

Product Code. 30283805

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DWIWNQMHID EEKNTSLPHH VGKIKSSVSR KNAKYLLKGE YVGKVFRVDA ETGDVFAIER LDRENISEYH LTAVIVDKDT GENLETPSSF TIKVHDVNDN WPVFTHRLFN ASVPESSAVG TSVISVTAVD ADDPTVGDHA SVMYQILKGK EYFAIDNSGR IITITKSLDR EKQARYEIVV EARDAQGLRG DSGTATVLVT LQDINDNFPF FTQTKYTFVV PEDTRVGTSV GSLFVEDPDE PQNRMTKYSI LRGDYQDAFT IETNPAHNEG IIKPMKPLDY EYIQQYSFIV EATDPTIDLR YMSPPAGNRA QVIINITDVD EPPIFQQPFY HFQLKENQKK PLIGTVLAMD PDAARHSIGY SIRRTSDKGQ FFRVTKKGDI YNEKELDREV YPWYNLTVEA KELDSTGTPT GKESIVQVHI EVLDENDNAP EFAKPYQPKV CENAVHGQLV LQISAIDKDI TPRNVKFKFI LNTENNFTLT DNHDNTANIT VKYGQFDREH TKVHFLPVVI SDNGMPSRTG TSTLTVAVCK CNEQGEFTFC EDMAAQ.

VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VE-cadherin
Recombinant Monoclonal
0.13 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P33151
CDH5
Recombinant fusion protein containing a sequence corresponding to amino acids 48-593 of human VE Cadherin (NP_001786.2).
100 μL
Primary
Human
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot
9W5E3
Unconjugated
CDH5
7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD
Rabbit
Protein A
RUO
1003
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.