missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VE-cadherin Monoclonal Antibody (3D5C7)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA517050

Product Code. 15357970

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-17050 targets CDH5 in FACS, indirect ELISA, and WB applications and shows reactivity with Human and Rat samples. The MA5-17050 immunogen is purified recombinant fragment of human CDH5 (amino acids: 29-223 ) expressed in E. Coli. MA5-17050 detects CDH5 which has a predicted molecular weight of approximately 87.5kDa.

VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VE-cadherin
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P33151
CDH5
Purified recombinant fragment of human CDH5 (amino acids: 29-223 ) expressed in E. Coli.
100 μg
Primary
Human, Rat
Antibody
IgG1
ELISA, Flow Cytometry, Western Blot
3D5C7
Unconjugated
CDH5
7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD
Mouse
Protein G
RUO
1003, 307618
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.