missing translation for 'onlineSavingsMsg'
Learn More

VE-821, MedChemExpress

Product Code. 16448747
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448747 5 mg 5mg
16426148 10 mg 10mg
16406536 100 mg 100mg
16402287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16448747 Supplier Medchem Express Supplier No. HY14731
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress VE-821 is a potent ATP-competitive inhibitor of ATR with Ki/IC50 of 13 nM/26 nM.

VE-821 is a potent ATP-competitive inhibitor of ATR with Ki/IC50 of 13 nM/26 nM.

Specifications

Chemical Name or Material VE-821
CAS 1232410-49-9
Color Green
Purity Grade Notes Research
Molecular Formula C18H16N4O3S
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (135.72 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(NC1=CC=CC=C1)C2=NC(C3=CC=C(C=C3)S(=O)(C)=O)=CN=C2N
Molecular Weight (g/mol) 368.41
Formula Weight 368.41
Percent Purity 98.94%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.