missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDR/NR1I1/Vitamin D Receptor Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-794
This item is not returnable.
View return policy
Description
VDR/NR1I1/Vitamin D Receptor Polyclonal specifically detects VDR/NR1I1/Vitamin D Receptor in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| VDR/NR1I1/Vitamin D Receptor | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| NR1I1Nuclear receptor subfamily 1 group I member 11,25-dihydroxyvitamin D3 receptor, vitamin D1,25- dihydroxyvitamin D3 receptor, vitamin D3 receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7421 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1.0 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_000367.1 | |
| VDR | |
| Peptide with sequence CGNQDYKYRVSD, from the internal region of the protein sequence according to NP_000367.1; NP_001017535.1. | |
| 0.1 mg | |
| Primary | |
| Both transcript variants (NP_000367; NP_001017535) encode the same protein. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction