missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VDR Monoclonal Antibody (9A7gamma.E10.E4)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA514617
This item is not returnable.
View return policy
Description
MA5-14617 targets Vitamin D Receptor in GS and WB applications and shows reactivity with Amphibian, Chicken, Fish, Hamster, Human, mouse, Porcine, and Rat samples. The MA5-14617 immunogen is vitamin D receptor protein from chicken intestine.
Steroid receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. The 1,25-dihydroxy-vitamin D3 receptor (VDR) belongs to the superfamily of steroid hormone receptors which includes estrogen, progesterone, glucocorticoid, androgen, and thyroid hormone receptors. Vitamin D modulates calcium and phosphorus homeostasis, bone remodeling, cell growth regulation, and differentiation. Studies have found VDR in the intestine, bone, kidney, epidermis, and cells of the endocrine immune system. Repression of T-cell proliferation and interleukin-2 (IL-2) gene expression, for example, occurs when VDR binds within the IL-2 enhancer. Formation of VDR/retinoic X receptor (RXR) heterodimers in the presence of intracellular 1,25(OH)2D3 has been shown to interfere with assembly of nuclear factor of activated T-cells (NFATp)/Fos/Jun/DNA complex and subsequent IL-2 gene transcription. The corresponding gene for the vitamin D receptor is NR1I1.
Specifications
| VDR | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| A3RGC1, O42392, P11473, P13053, P48281 | |
| VDR | |
| Vitamin D receptor protein from chicken intestine. | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat, Hamster, Pig, Chicken, Amphibian, Fish | |
| Antibody | |
| IgG2b |
| Gel Shift, Western Blot, Immunocytochemistry | |
| 9A7gamma.E10.E4 | |
| Unconjugated | |
| VDR | |
| 1,25-dihydroxyvitamin D3 receptor; HGNC:12679; I79_025795; Nr1i1; Nuclear receptor subfamily 1 group I member 1; PPP1R163; protein phosphatase 1, regulatory subunit 163; vdr; vdr.S; vdr-A; vitamin D (1,25- dihydroxyvitamin D3) receptor; vitamin D (1,25- dihydroxyvitamin D3) receptor S homeolog; vitamin D (1,25-dihydroxyvitamin D3) receptor; vitamin D nuclear receptor variant 1; vitamin D receptor; vitamin D receptor protein; vitamin D receptor S homeolog; Vitamin D3 receptor; XELAEV_18015993mg; xVDR | |
| Rat | |
| Protein G | |
| RUO | |
| 100767048, 22337, 24873, 395988, 396628, 7421 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction