missing translation for 'onlineSavingsMsg'
Learn More

VDP p115 Antibody (3F4), Novus Biologicals™

Product Code. 18329689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329689 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329689 Supplier Novus Biologicals Supplier No. H00008615M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VDP p115 Monoclonal antibody specifically detects VDP p115 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen VDP p115
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 3F4
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH32654
Gene Alias p115, Transcytosis-associated protein, USO1 homolog, vesicle docking protein (yeast), USO1 vesicle docking protein homolog (yeast), vesicle docking protein, vesicle docking protein p115
Host Species Mouse
Immunogen USO1 (AAH32654, 1 a.a. ~ 961 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MNFLRGVMGGQSAGPQHTEAETIQKLCDRVASSTLLDDRRNAVRALKSLSKKYRLEVGIQAMEHLIHVLQTDRSDSEIIGYALDTLYNIISNEEEEVEENSTRQSEDLGSQFTEIFIKQQENVTLLLSLLEEFDFHVRWPGVKLLTSLLKQLGPQVQQIILVSPMGVSRLMDLLADSREVIRNDGVLLLQALTRSNGAIQKIVAFENAFERLLDIISEEGNSDGGIVVEDCLILLQNLLKNNNSNQNFFKEGSYIQRMKPWFEVGDENSGWSAQKVTNLHLMLQLVRVLVSPTNPPGATSSCQKAMFQCGLLQQLCTILMATGVPADILTETINTVSEVIRGCQVNQDYFASVNAPSNPPRPAIVVLLMSMVNERQPFVLRCAVLYCFQCFLYKNQKGQGEIVSTLLPSTIDATGNSVSAGQLLCGGLFSTDSLSNWCAAVALAHALQENATQKEQLLRVQLATSIGNPPVSLLQQCTNILSQGSKIQTRVGLLMLLCTWLSNCPIAVTHFLHNSANVPFLTGQIAENLGEEEQLVQGLCALLLGISIYFNDNSLESYMKEKLKQLIEKRIGKENFIEKLGFISKHELYSRASQKPQPNFPSPEYMIFDHEFTKLVKELEGVITKAIYKSSEEDKKEEEVKKTLEQHDNIVTHYKNMIREQDLQLEELRQQVSTLKCQNEQLQTAVTQQVSQIQQHKDQYNLLKIQLGKDNQHQGSYSEGAQMNGIQPEEIGRLREEIEELKRNQELLQSQLTEKDSMIENMKSSQTSGTNEQSSAIVSARDSEQVAELKQELATLKSQLNSQSVEITKLQTEKQELLQKTEAFAKSVEVQGETETIIATKTTDVEGRLSALLQETKELKNEIKALSEERTAIKEQLDSSNSTIAILQTEKDKLELEITDSKKEQDDLLVLLADQDQKILSLKNKLKDLGHPVEEEDELESGDQEDEDDESEDPGKDLDHI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8615
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.