missing translation for 'onlineSavingsMsg'
Learn More

VDAC3 Antibody (1C6), Novus Biologicals™

Product Code. 18352579 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352579 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352579 Supplier Novus Biologicals Supplier No. H00007419M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VDAC3 Monoclonal antibody specifically detects VDAC3 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen VDAC3
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1C6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH56870
Gene Alias HD-VDAC3, hVDAC3, Outer mitochondrial membrane protein porin 3, VDAC-3, voltage-dependent anion channel 3, voltage-dependent anion-selective channel protein 3
Host Species Mouse
Immunogen VDAC3 (AAH56870, 1 a.a. ~ 283 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MCNTPTYCDLGKAAKDVFNKGYGFGMVKIDLKTKSCSGVEFSTSGHAYTDTGKASGNLETKYKVCNYGLTFTQKWNTDNTLGTEISWENKLAEGLKLTLDTIFVPNTGKKSGKLKASYKRDCFSVGSNVDIDFSGPTIYGWAVLAFEGWLAGYQMSFDTAKSKLSQNNFALGYKAADFQLHTHVNDGTEFGGSIYQKVNEKIETSINLAWTAGSNNTRFGIAAKYMLDCRTSLSAKVNNASLIGLGYTQTLRPGVKLTLSALIDGKNFSAGGHKVGLGFELEA
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7419
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.