missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDAC1 Rabbit anti-Human, Mouse, Rat, Clone: S05-7F2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-14982-100UL
This item is not returnable.
View return policy
Description
VDAC1 Monoclonal antibody specifically detects VDAC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| VDAC1 | |
| Monoclonal | |
| Unconjugated | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S05-7F2 | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:20-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| hVDAC1, MGC111064, Outer mitochondrial membrane protein porin 1, Plasmalemmal porin, PORIN, Porin 31HL, Porin 31HM, PORIN-31-HL, VDAC, VDAC-1, voltage-dependent anion channel 1, voltage-dependent anion-selective channel protein 1 | |
| A synthetic peptide of human VDAC1 (Uniprot#: P21796) | |
| 100 μg | |
| Cellular Markers, Mitochondrial Markers | |
| 7416 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction