missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDAC1 Rabbit anti-Human, Mouse, Rat, Bovine, Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-45921-0.025MG
This item is not returnable.
View return policy
Description
VDAC1 Polyclonal specifically detects VDAC1 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| VDAC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
| hVDAC1, MGC111064, Outer mitochondrial membrane protein porin 1, Plasmalemmal porin, PORIN, Porin 31HL, Porin 31HM, PORIN-31-HL, VDAC, VDAC-1, voltage-dependent anion channel 1, voltage-dependent anion-selective channel protein 1 | |
| Rabbit | |
| 31 kDa | |
| 0.025 mg | |
| Cellular Markers, Mitochondrial Markers | |
| 7416 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 2.5 ug/mL | |
| P21796 | |
| VDAC1 | |
| Synthetic peptide surrounding amino acid 189 of human VDAC. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, Rat, Human, Bovine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction