missing translation for 'onlineSavingsMsg'
Learn More

VD3-d6, MedChemExpress

Product Code. 16469047
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469047 5 mg 5mg
16431509 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469047 Supplier Medchem Express Supplier No. HY15331

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VD3-d6 (Vitamin D3-26,26,26,27,27,27-d6) is the deuterium labeled Vitamin D3; tools for determination of Vitamin D3 metabolites in human serum.

VD3-d6 (Vitamin D3-26,26,26,27,27,27-d6) is the deuterium labeled Vitamin D3

Specifications

Chemical Name or Material VD3-d6
CAS 118584-54-6
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C27H38D6O
Color Light Yellow
Synonym Vitamin D3-26,26,26,27,27,27-d6
Solubility Information DMSO : 100 mg/mL (255.97 mM; Need ultrasonic)
SMILES O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCCC(C([2H])([2H])[2H])C([2H])([2H])[2H])C)([H])[C@]3(C)CCC\2
Industry Type Neuroscience-Neuromodulation
Molecular Weight (g/mol) 390.67
Quantity 5 mg
Formula Weight 390.67
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.