missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VCX3A Monoclonal antibody specifically detects VCX3A in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Specifications
| Antigen | VCX3A |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 6A3 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:100 to 1:2000, Sandwich ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_057463 |
| Gene Alias | Variable charge protein on X with eight repeats, variable charge X-linked protein 3, variable charge, X-linked 3A, Variably charged protein X-A, variably charged X-A, VCX3, VCX-8R, VCX8RMGC125730, VCX-8rvariable charge, X-linked 3, VCX-AMGC118976, VCXAMGC125796 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?