missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VCP Recombinant Rabbit Monoclonal Antibody (HL3018)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556911
This item is not returnable.
View return policy
Description
VCP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.
Specifications
| VCP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P46462, P55072, Q01853, Q7ZU99 | |
| VCP | |
| Recombinant fragment of human VCP. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL3018 | |
| Unconjugated | |
| VCP | |
| 15S Mg(2+)-ATPase p97 subunit; 3110001E05; AAA ATPase p97; ALS14; CDC48; cell division cycle gene CDC48; CMT2Y; epididymis luminal protein 220; epididymis secretory protein Li 70; HEL-220; HEL-S-70; homolog of yeast cdc48; IBMPFD; IBMPFD1; Inv protein; or HEL-S-70; p97; p97 atpase; p97/VCP; Protein CDC48; si:ch211-113n10.2; TER ATPase; TERA; transitional endoplasmic reticulum ATPase; valosin containing protein; valosin-containing protein; Vcp; wu:fd16d05; wu:fj63d11; yeast Cdc48p homolog | |
| Rabbit | |
| Protein A | |
| RUO | |
| 116643, 269523, 327197, 7415 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction