missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VCP 171
GREENER_CHOICE

Product Code. 18779916
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18779916 10 mg 10mg
18759886 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18779916 Supplier Tocris Bioscience™ Supplier No. 6261/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulator (PAM) of A1 receptors; also allosteric partial agonist in absence of orthosteric agonist

VCP 171 is an adenosine A1 receptor positive allosteric modulator (PAM) (pKB = 5.65, binding cooperativity with NECA 0.68). In absence of orthosteric agonist, VCP171 behaves as an allosteric partial agonist, inhibiting cAMP activity. Inhibits AMPA receptor-mediated evoked excitatory postsynaptic current (eEPSC) amplitude in lamina I and II neurons in a rat partial nerve-injury model of neuropathic pain.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [2-Amino-4-[3-(trifluoromethyl)phenyl]-3-thienyl]phenylmethanone
CAS 1018830-99-3
Quantity 10 mg
Target Adenosine A1 Receptor Modulators
Molecular Formula C18H12F3NOS
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.