missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCIP135/VCPIP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94222-0.1ml
This item is not returnable.
View return policy
Description
VCIP135/VCPIP1 Polyclonal antibody specifically detects VCIP135/VCPIP1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| VCIP135/VCPIP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp686G038, DUBA3, EC 3.4.22.-, FLJ23132, KIAA1850FLJ60694, valosin containing protein (p97)/p47 complex interacting protein 1, valosin-containing protein (p97)/p47 complex-interacting protein p135, Valosin-containing protein p97/p47 complex-interacting protein 1, Valosin-containing protein p97/p47 complex-interacting protein p135, VCIP135deubiquitinating protein VCIP135, VCPIP1 VCIP135 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 923-1222 of human VCPIP1 (NP_079330.2). VFYSIMKKTMGMADGKHCTFPHLPGKTFVYNASEDRLELCVDAAGHFPIGPDVEDLVKEAVSQVRAEATTRSRESSPSHGLLKLGSGGVVKKKSEQLHNVTAFQGKGHSLGTASGNPHLDPRARETSVVRKHNTGTDFSNSSTKTEPSVFTASSSNSELIRIAPGVVTMRDGRQLDPDLVEAQRKKLQEMVSSIQASMDRHLRDQSTEQSPSDLPQRKTEVVSSSAKSGSLQTGLPESFPLTGGTENLNTETTDGCVADALGAAFATRSKAQRGNSVEELEEMDSQDAEMTNTTEPMDHS | |
| 0.1 mL | |
| Cell Biology | |
| 80124 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction