missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vcan, Mouse anti-Mouse, PCP, Clone: S351-23, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB18515.100ug
This item is not returnable.
View return policy
Specifications
| Vcan | |
| Monoclonal | |
| PerCP | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| 5430420N07Rik/9430051N09/Cspg2/DPEAAE/NG2/PG-M/PG-M(V0)/PG-M(V1)/hdf | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| S351-23 | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| versican | |
| Vcan | |
| Recombinant protein corresponding to amino acids 362-585 at glycosaminoglycan alpha domain of mouse Vcan core protein. | |
| RUO | |
| 13003 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction