Learn More
VCAM-1 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30958-1-AP-20UL
Description
The VCAM-1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse VCAM-1. This antibody recognizes mouse, rat antigen. The VCAM-1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| VCAM-1 | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| CD106, Vcam 1, Vcam1 | |
| 20 μL | |
| 22329 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |