Learn More
Invitrogen™ VCAM-1 Monoclonal Antibody (6G9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515636
Description
MA5-15636 targets VCAM1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15636 immunogen is purified recombinant fragment of human VCAM1 expressed in E. Coli. MA5-15636 detects VCAM1 which has a predicted molecular weight of approximately 81kDa.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
Specifications
| VCAM-1 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| P19320 | |
| VCAM1 | |
| Purified recombinant fragment of human VCAM1 expressed in E. Coli. | |
| RUO | |
| 7412 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6G9 | |
| Unconjugated | |
| VCAM1 | |
| adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.