missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38587-100ul
This item is not returnable.
View return policy
Description
VBP1 Polyclonal antibody specifically detects VBP1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| VBP1 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, ELISA | |
| HIBBJ46, PFD3, PFDN3VHL-binding protein 1, prefoldin 3, prefoldin subunit 3, VBP-1, VHL binding protein-1, von Hippel-Lindau binding protein 1, Von Hippel-Lindau-binding protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-197 of human VBP1 (NP_003363.1).,, Sequence:, MAAVKDSCGKGEMATGNGRRLHLGIPEAVFVEDVDSFMKQPGNETADTVLKKLDEQYQKYKFMELNLAQKKRRLKGQIPEIKQTLEILKYMQKKKESTNSMETRFLLADNLYCKASVPPTDKVCLWLGANVMLEYDIDEAQALLEKNLSTATKNLDSLEEDLDFLRDQFTTTEVNMARVYNWDVKRRNKDDSTKNKA | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 7411 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction