Learn More
VAX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564931
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: RRAESGGGGGR CGDRSGAGDL RADGGGHSPT EVAGTSASSP AGSRESGADS DGQPGPGEAD HCRRILVRDA KGTIREIVLP KGLDLDRPKR T Highest antigen sequence identity to the following orthologs - mouse 79%, rat 78%.
This gene encodes a homeobox protein and is almost exclusively expressed in the ventral portion of the retina during development. In mouse studies, this gene was found to be required for the correct formation of the optic fissure and other aspects of retinal development.Specifications
VAX2 | |
Polyclonal | |
Unconjugated | |
VAX2 | |
DRES93; VAX2; Ventral anterior homeobox 2; ventral anterior homeobox containing gene 2; ventral retina homeodomain protein; Vex | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25806 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UIW0 | |
VAX2 | |
Recombinant Human VAX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only