missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VAX1 Recombinant Mouse Monoclonal Antibody (K29066_16C2)

Product Code. 30829328
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30829328 100 μL -
30828915 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829328 Supplier Invitrogen™ Supplier No. MA564459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: RCHSDAEAAR VSKNAHKESR ESKGAEGNLP AAFLKEPQGA FSASGAAEDC NKSKSNSAAD PDYCRRILVR DAKGSIREII LPKGLDLDRP KRTRTSFTAE QLYRLEMEFQ RCQYVVGRER TELARQLNLS ETQVKVWFQN RRTKQKKDQG KDSELRSVVS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%, Pig - 99%, Cynomolgus monkey - 99%.

This gene encodes a homeo-domain containing protein from a class of homeobox transcription factors which are conserved in vertebrates. Genes of this family are involved in the regulation of body development and morphogenesis. The most conserved genes, called HOX genes are found in special gene clusters. This gene belongs to the VAX subfamily and lies in the vicinity of the EMX homeobox gene family. Another member of VAX family is located on chromosome 2. The encoded protein may play an important role in the development of anterior ventral forebrain and visual system. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAX1
Applications Immunoprecipitation
Classification Recombinant Monoclonal
Clone K29066_16C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene VAX1
Gene Accession No. Q5SQQ9
Gene Alias homeodomain protein Vax1; MCOPS11; VAX1; Ventral anterior homeobox 1; ventral anterior homeobox containing gene 1
Gene Symbols VAX1
Host Species Mouse
Immunogen Recombinant human VAX1 (aa 11-170).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11023
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.