missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAV3 Recombinant Rabbit Monoclonal Antibody (JE33-02)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556548
This item is not returnable.
View return policy
Description
VAV3 Recombinant Monoclonal Antibody for Western Blot
This gene is a member of the VAV gene family. The VAV proteins are guanine nucleotide exchange factors (GEFs) for Rho family GTPases that activate pathways leading to actin cytoskeletal rearrangements and transcriptional alterations. This gene product acts as a GEF preferentially for RhoG, RhoA, and to a lesser extent, RAC1, and it associates maximally with the nucleotide-free states of these GTPases. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| VAV3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9UKW4 | |
| VAV3 | |
| Recombinant protein within human vav3 aa 748-847/847. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| JE33-02 | |
| Unconjugated | |
| VAV3 | |
| A530094I06Rik; AA986410; FLJ40431; guanine nucleotide exchange factor VAV3; Idd18.1; RGD1565941; vav 3 guanine nucleotide exchange factor; vav 3 oncogene; vav guanine nucleotide exchange factor 3; Vav3; VAV-3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10451, 295378 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction