missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VAV3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16537831
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16537831 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16537831 Supplier Invitrogen™ Supplier No. PA590428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DNCGRVNSGE QGTLKLPEKR TNGLRRTPKQ VDPGLPKMQV IRNYSGTPPP ALHEGPPLQL QAGDTVELLK GDAHSLFWQG RNLASGEVGF FPSDAVKPCP CVPKPVDYSC QPWYAGAMER L; Positive Samples: Jurkat, Mouse brain, Mouse lung, Mouse spleen.

This gene is a member of the VAV gene family. The VAV proteins are guanine nucleotide exchange factors (GEFs) for Rho family GTPases that activate pathways leading to actin cytoskeletal rearrangements and transcriptional alterations. This gene product acts as a GEF preferentially for RhoG, RhoA, and to a lesser extent, RAC1, and it associates maximally with the nucleotide-free states of these GTPases. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAV3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene VAV3
Gene Accession No. Q9R0C8, Q9UKW4
Gene Alias A530094I06Rik; AA986410; FLJ40431; guanine nucleotide exchange factor VAV3; Idd18.1; RGD1565941; vav 3 guanine nucleotide exchange factor; vav 3 oncogene; vav guanine nucleotide exchange factor 3; Vav3; VAV-3
Gene Symbols VAV3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 560-680 of human VAV3 (NP_0061044).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10451, 57257
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.