Learn More
Invitrogen™ VAV2 Recombinant Rabbit Monoclonal Antibody (SC68-03)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532245
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| VAV2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P52735, Q60992 | |
| Vav2 | |
| Synthetic peptide within Human VAV2 aa 146-194. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| SC68-03 | |
| Unconjugated | |
| Vav2 | |
| 2810040F13Rik; AI847175; Guanine nucleotide exchange factor VAV2; Oncogene VAV2; vav 2 guanine nucleotide exchange factor; vav 2 oncogene; vav guanine nucleotide exchange factor 2; VAV2; VAV-2; Vav2 oncogene | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22325, 296603, 7410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.