missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAV2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA19115
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| VAV2 | |
| Polyclonal | |
| Unconjugated | |
| Vav2 | |
| 2810040F13Rik; AI847175; Guanine nucleotide exchange factor VAV2; Oncogene VAV2; vav 2 guanine nucleotide exchange factor; vav 2 oncogene; vav guanine nucleotide exchange factor 2; VAV2; VAV-2; Vav2 oncogene | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 7410 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P52735 | |
| Vav2 | |
| Internal sequence corresponding to amino acids C-ETEAKYYRTLEDIEC (aa 208-221). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction