missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ VAV2 Antibody, Novus Biologicals™

Sheep Polyclonal Antibody

£162.00 - £386.00

Specifications

Antigen VAV2
Dilution Western Blot : 1:5000, Immunoprecipitation
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30067076
View Documents
Novus Biologicals™
NBP3-30790-250ug
250 μg
£386.00
250µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30067078
View Documents
Novus Biologicals™
NBP3-30790-50ug
50 μg
£162.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VAV2 Polyclonal antibody specifically detects VAV2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications

Specifications

VAV2
Immunoprecipitation, Western Blot
Unconjugated
Sheep
Signal Transduction
Phosphate buffered saline
7410
IgG
Ammonium sulfate precipitation
Western Blot : 1:5000, Immunoprecipitation
Polyclonal
Purified
RUO
Human
guanine nucleotide exchange factor VAV2, oncogene VAV2, Protein vav-2, vav 2 guanine nucleotide exchange factor, vav 2 oncogene, VAV-2
A his tagged fusion protein VAV2 corresponding to amino acid sequence 578 - 878 and having a molecular weight of 100kD
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.