missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAV1 Polyclonal Antibody
Description
Recommended positive controls: Jurkat, NCI-H929, BCL-1. Predicted reactivity: Mouse (92%), Rat (92%), Pig (96%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The VAV1 gene, located on chromosome 19p13.2, encodes for a protein that functions as a guanine nucleotide exchange factor (GEF). Structurally, the VAV1 protein contains several functional domains, including Dbl homology (DH) and pleckstrin homology (PH) domains, which facilitate its role in exchange activity, and SH3 and SH2 domains, critical for signal transduction. VAV1 is primarily expressed in hematopoietic cells and plays a crucial role in the regulation of immune responses, particularly in T-cell receptor signaling. It influences actin cytoskeleton reorganization, cell proliferation, and differentiation, making it vital for proper immune system function.
Specifications
Specifications
| Antigen | VAV1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.49 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 |
| Gene | VAV1 |
| Gene Accession No. | P15498, P27870 |
| Gene Alias | oncogene vav; p95; p95vav; Proto-oncogene vav; VAV; vav 1 guanine nucleotide exchange factor; vav 1 oncogene; vav guanine nucleotide exchange factor 1; vav oncogene; vav proto-oncogene; VAV1; vav-T |
| Gene Symbols | VAV1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?