missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAV1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517834
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Vav2, Vav3 or CelVav.
This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14.7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| VAV1 | |
| Polyclonal | |
| Unconjugated | |
| VAV1 | |
| oncogene vav; p95; p95vav; Proto-oncogene vav; VAV; vav 1 guanine nucleotide exchange factor; vav 1 oncogene; vav guanine nucleotide exchange factor 1; vav oncogene; vav proto-oncogene; VAV1; vav-T | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22324, 25156, 7409 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 22 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P15498, P27870, P54100 | |
| VAV1 | |
| Synthetic peptide corresponding to residues surrounding Tyr174 of human Vav. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction