missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAV1 Monoclonal Antibody (GT557)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531488
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (92%), Rat (92%), Pig (96%), Bovine (96%). Positive Control: Jurkat, NCI-H929. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14.7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| VAV1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P15498, P27870 | |
| VAV1 | |
| Recombinant protein encompassing a sequence within the center region of human VAV1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT557 | |
| Unconjugated | |
| VAV1 | |
| oncogene vav; p95; p95vav; Proto-oncogene vav; VAV; vav 1 guanine nucleotide exchange factor; vav 1 oncogene; vav guanine nucleotide exchange factor 1; vav oncogene; vav proto-oncogene; VAV1; vav-T | |
| Mouse | |
| Protein G | |
| RUO | |
| 22324, 7409 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction