missing translation for 'onlineSavingsMsg'
Learn More

Vatinoxan hydrochloride, MedChemExpress

Product Code. 16490148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. Quantity unitSize
16490148 5 mg 5mg
16477688 10 mg 10mg
16403636 25 mg 25mg
3 options
This item is not returnable. View return policy

Product Code. 16490148

Brand: Medchem Express HY19057A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vatinoxan hydrochloride (MK-467 hydrochloride;L-659066 hydrochloride) is a peripheral α2 adrenergic receptor antagonist.

Vatinoxan hydrochloride (MK-467 hydrochloride

Specifications

Chemical Name or Material Vatinoxan hydrochloride
CAS 130466-38-5
Color White
Purity Grade Notes Research
Molecular Formula C20H27ClN4O4S
Quantity 5 mg
Synonym MK-467 hydrochloride L-659066 hydrochloride
Solubility Information H2O : ≥ 133 mg/mL (292.33 mM)
SMILES [H]Cl.CS(=O)(NCCN([C@]1(CN2)CCN3CCC4=C(OC5=CC=CC=C54)[C@]3([H])C1)C2=O)=O
Molecular Weight (g/mol) 454.97
Formula Weight 454.97
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.