missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Vatalanib succinate
GREENER_CHOICE

Product Code. p-7110874
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18780919

Brand: Tocris Bioscience™ 5680/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent VEGFR inhibitor; also aromatase inhibitor

Vatalanib succinate is a potent VEGFR inhibitor (IC50 values are 37 and 77 nM for VEGFR-2 and -1, respectively). Inhibits proliferation, migration and survival of HUVECs in vitro and inhibits growth, vascularization and metastasis of tumors expressing VEGFR in mouse models. Also inhibits PDGFR-β, c-Kit and c-Fms. Potent aromatase inhibitor (IC50 = 50 nM). Orally available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4-Chlorophenyl)-4-(4-pyridinylmethyl)-1-phthalazinamine succinate
CAS 212142-18-2
Synonym CGP 79787D, PTK787/ZK222584
Quantity 50 mg
Target VEGF Receptor Inhibitors
Molecular Formula C20H15ClN4.C4H6O4
Formula Weight 464.9
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.