missing translation for 'onlineSavingsMsg'
Learn More

Vatalanib dihydrochloride, MedChemExpress

Product Code. 16404928
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16404928

Brand: Medchem Express HY12018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vatalanib dihydrochloride (PTK787 dihydrochloride) is an inhibitor of VEGFR2/KDR with IC50 of 37 nM.

Vatalanib dihydrochloride (PTK787 dihydrochloride) is an inhibitor of VEGFR2/KDR with IC50 of 37 nM.

Specifications

Chemical Name or Material Vatalanib dihydrochloride
Physical Form Solid
CAS 212141-51-0
Color Light Nude
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H17Cl3N4
For Use With (Application) Cancer-Kinase/protease
Synonym PTK787 dihydrochloride CGP-797870 dihydrochloride ZK-222584 dihydrochloride
Solubility Information DMSO : 50 mg/mL (119.12 mM; ultrasonic and warming and heat to 80°C) ∣H2O : 50 mg/mL (119.12 mM; Need ultrasonic)
SMILES ClC1=CC=C(C=C1)NC2=NN=C(C3=C2C=CC=C3)CC4=CC=NC=C4.[H]Cl.[H]Cl
Molecular Weight (g/mol) 419.73
Formula Weight 419.73
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.