Learn More
VAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543777
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PPAPGPGQLT LRLRACGLNF ADLMARQGLY DRLPPLPVTP GMEGAGVVIA Sequence homology: Cow: 93%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rat: 100%; Zebrafish: 86%
VAT1 Polyclonal antibody specifically detects VAT1 in Human samples. It is validated for Western Blot.Specifications
VAT1 | |
Polyclonal | |
Unconjugated | |
VAT1 | |
FLJ20230; membrane protein of cholinergic synaptic vesicles; mitofusin-binding protein; Protein MIB; si:dz163l24.2; solute carrier family 18 (vesicular monoamine), member 1; synaptic vesicle membrane protein VAT-1 homolog; VAT1; Vat-1; vat1 protein; VATI; vesicle amine transport 1; vesicle amine transport protein 1 homolog (T californica); vesicle amine transport protein 1 homolog (T. californica) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10493 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q99536 | |
VAT1 | |
Synthetic peptide directed towards the N-terminal of human VAT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |