missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560941
This item is not returnable.
View return policy
Description
Immunogen sequence: GDRVMVLNRS GMWQEEVTVP SVQTFLIPEA MTFEEAAALL VNYITAYMVL FDFGNLQPGH SVLVH Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Synaptic vesicles are responsible for regulating the storage and release of neurotransmitters in the nerve terminal. The protein encoded by this gene is an abundant integral membrane protein of cholinergic synaptic vesicles and is thought to be involved in vesicular transport. It belongs to the quinone oxidoreductase subfamily of zinc-containing alcohol dehydrogenase proteins.
Specifications
| VAT1 | |
| Polyclonal | |
| Unconjugated | |
| VAT1 | |
| FLJ20230; membrane protein of cholinergic synaptic vesicles; mitofusin-binding protein; Protein MIB; si:dz163l24.2; solute carrier family 18 (vesicular monoamine), member 1; synaptic vesicle membrane protein VAT-1 homolog; VAT1; Vat-1; vat1 protein; VATI; vesicle amine transport 1; vesicle amine transport protein 1 homolog (T californica); vesicle amine transport protein 1 homolog (T. californica) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10493 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99536 | |
| VAT1 | |
| Recombinant protein corresponding to Human VAT1. Recombinant protein control fragment (Product #RP-88743). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction