missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VASP Recombinant Rabbit Monoclonal Antibody (23GB4715), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | VASP |
|---|---|
| Clone | 23GB4715 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
30088653
![]() |
Invitrogen™
MA553794 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087642
|
Invitrogen™
MA553795 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
VASP Recombinant Monoclonal antibody specifically detects VASP in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotEspecificaciones
| VASP | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P50552 | |
| 7408 | |
| IgG | |
| Affinity Chromatography | |
| Antibody |
| 23GB4715 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Enah/Vasp-like; I79_012660; Unknown (protein for MGC:127789); vasodilator stimulated phosphoprotein; vasodilator-stimulated phosphoprotein; vasodilator-stimulated phosphoprotein (VASP); VASP | |
| A synthesized peptide derived from human VASP (44-94AA). | |
| Primary | |
| -20°C | |
| VASP |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto